1-Pentanesulphonic acid sodium salt monohydrate 98+% for HPLC

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

423200250 423201000
ACRO423200250EA 85.8 GBP
ACRO423200250 ACRO423201000
1-Pentanesulphonic acid sodium salt monohydrate 98+% for HPLC
1-Pentanesulphonic acid sodium salt monohydrate
Formula: C₅H₁₁NaO₃S·1H₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007541
CAS Number: 207605-40-1
EINECS: 245-208-4

Order Now

Specification Test Results

Appearance Colorless to white Powder or crystals or flakes
Infrared spectrum Conforms
UV at 260 nm A: ≤0.01
Titration after Ion exchange ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR