Palladium 10% on activated carbon wetted with ca. 50% water, unreduced

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

422980100 422980250 422980010 422981000
ACRO422980100EA 301 GBP
ACRO422980100 ACRO422980250 ACRO422980010 ACRO422981000
Palladium 10% on activated carbon wetted with ca. 50% water, unreduced
Palladium

Reagent 50% wet with water for safety

Formula: Pd
Melting Pt: 1555 °C
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now

Specification Test Results

Appearance Black Powder
Assay 9.7 to 10.3 % (Pd)
Loss on drying 45 % to 55 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR