Wright's stain, pure_certified for microscopical staining

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue

422640100 422640250 422641000
ACRO422640100EA 27.1 GBP
ACRO422640100 ACRO422640250 ACRO422641000
Wright's stain, pure_certified for microscopical staining
Wright's stain
Storage Temperature: Ambient
MDL Number: MFCD00082143
CAS Number: 68988-92-1
EINECS: 273-541-5

Order Now

Specification Test Results

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR