N-(4-Vinylbenzyl)-N,N-dimethylamine 90% stabilised, Technical Grade

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

422550500 422550050
ACRO422550500EA 316 GBP
ACRO422550500 ACRO422550050
N-(4-Vinylbenzyl)-N,N-dimethylamine 90% stabilised, Technical Grade
N-(4-Vinylbenzyl)-N,N-dimethylamine

CAS No.: 2245-52-5

Formula: C₁₁H₁₅N
Boiling Pt: 72…74 °C (1 mmHg)
Density: 0.92 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00078270
CAS Number: 2245-52-5

Order Now

Specification Test Results

Appearance Clear yellow to orange to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5360 to 1.5400 (20°C, 589 nm)
GC ≥88 %
Stabilizer 0.1 % Hydroquinone


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR