Trichloroethylene 99.6% stabilised ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 1,1,2-Trichloroethene, Trichloroethene, 1,1,2-Trichloroethylene

421525000 421520025
ACRO421525000EA 28.4 GBP
ACRO421525000 ACRO421520025
Trichloroethylene 99.6% stabilised ACS
Trichloroethylene
Formula: ClCH=CCl₂
MW: 131.39 g/mol
Boiling Pt: 86.7
Melting Pt: –85 °C
Density: 1.46 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000838
CAS Number: 79-01-6
UN: 1710
ADR: 6.1 III
Merck Index: 12,09769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ac57d7a9ea2e3097408c3559993eb33587aec09&vid=767d7d09973e747c5a2d07a4d46dffedfb559ceb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.02 % (Coulometric)
GC ≥99.5 %
Color scale ≤10 APHA
Stabilizer ≤1 % (Type of stabilizer mentioned on the coa)
Residue after evaporation ≤0.001 %
Free halogens Passes test
Heavy metals (as Pb) ≤1 ppm
Titrable acid ≤0.0001 meq/g
Titrable base ≤0.0003 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR