Tetrapropylammonium hydroxide 25% in water

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

420730250 420731000 420735000
ACRO420730250EA 26.3 GBP
ACRO420730250 ACRO420731000 ACRO420735000
Tetrapropylammonium hydroxide 25% in water
Tetrapropylammonium hydroxide
Formula: C₁₂H₂₉NO
Density: 1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00009360
CAS Number: 4499-86-9
EINECS: 224-800-6
UN: 3267
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration with HClO4 24 to 26 %
Melting point 114 to 118 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR