Tetramethylammonium hydroxide 25% in water

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

420521000 420520010 420520050
ACRO420521000EA 26.7 GBP
ACRO420521000 ACRO420520010 ACRO420520050
Tetramethylammonium hydroxide 25% in water
Tetramethylammonium hydroxide
Formula: C₄H₁₃NO
MW: 91.15 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –25 °C
Density: 1.014 g/cm³
Flash Pt: >95 °C
Storage Temperature: Ambient
MDL Number: MFCD00008280
CAS Number: 75-59-2
EINECS: 200-882-9
UN: 1835
ADR: 8 II
Merck Index: 14,09224

Order Now

Specification Test Results

Appearance Clear colorless to yellow Solution
Refractive index 1.3780 to 1.3840 (20°C, 589 nm)
Titration with HCl 23 to 27 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR