Tetramethylammonium hydroxide 10% in water

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

420511000 420515000 420510010
ACRO420511000EA 26 GBP
ACRO420511000 ACRO420515000 ACRO420510010
Tetramethylammonium hydroxide 10% in water
Tetramethylammonium hydroxide
Formula: C₄H₁₃NO
MW: 91.15 g/mol
Density: 1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00008280
CAS Number: 75-59-2
EINECS: 200-882-9
UN: 1835
ADR: 8 II
Merck Index: 14,09224

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration with HCl 9.5 to 10.5 %
Refractive index 1.6020 to 1.6050 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR