Tetrabutylammonium nitrate 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

420130250 420130050 420131000
ACRO420130250EA 72.9 GBP
ACRO420130250 ACRO420130050 ACRO420131000
Tetrabutylammonium nitrate 98%
Tetrabutylammonium nitrate
Formula: C₁₆H₃₆N₂O₃
Melting Pt: 116…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00043202
CAS Number: 1941-27-1
EINECS: 217-726-0
UN: 1479
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %
HPLC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR