Silver nitrate 99.5% for analysis

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

419361000 419365000 419360250
ACRO419361000EA 308 GBP
ACRO419361000 ACRO419365000 ACRO419360250
Silver nitrate 99.5% for analysis
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444
Melting Pt: 212 °C
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1 II
REACH: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration Argentometric ≥99.4 %
Chloride (Cl) ≤5 ppm
Cadmium (Cd) ≤1 ppm
Calcium (Ca) ≤10 ppm
Chromium (Cr) ≤0.2 ppm
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤0.4 ppm
Lead (Pb) ≤0.2 ppm
Manganese (Mn) ≤0.1 ppm
Nickel (Ni) ≤0.2 ppm
Palladium (Pd) ≤0.1 ppm
Sulfur (S) ≤20 ppm
Zinc (Zn) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR