Salicylic acid 99+% ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

419220020 419225000 419220100
ACRO419220020EA 127 GBP
ACRO419220020 ACRO419225000 ACRO419220100
Salicylic acid 99+% ACS
Salicylic acid
Formula: (HO)C₆H₄CO₂H
Boiling Pt: 211 °C (20 mmHg)
Melting Pt: 158…161 °C
Density: 1,443 g/cm³ (20 °C)
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00002439
CAS Number: 69-72-7
EINECS: 200-712-3
Merck Index: 13,08411
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af1f695fa92656a135b5e1742f1cca28f4d255fb&vid=8f3aff6fb9a70e5dd194b8bc46019af74e48e90a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 158.0 to 161.0 °C
HPLC ≥99.0 %
Residue after ignition ≤0.01 %
Substance darkened by H2SO4 passes test
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤30 ppm
Iron (Fe) ≤2 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR