1,10-Phenanthroline monohydrate ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 1,10-Phenanthroline hydrate

417120010 417120050 417120250 417121000
ACRO417120010EA 18.9 GBP
ACRO417120010 ACRO417120050 ACRO417120250 ACRO417121000
1,10-Phenanthroline monohydrate ACS
1,10-Phenanthroline monohydrate
Formula: C₁₂H₈N₂·1H₂O
MW: 198.22 g/mol
Melting Pt: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00149973
CAS Number: 5144-89-8
EINECS: 200-629-2
UN: 2811
ADR: 6.1 III
Merck Index: 13,07291
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba346b9f67fdda9afd4d48b893e6318564d604c&vid=2d08e04ee9525293a747b55cdcff8f66a6c4715a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or needles (May discolor to cream on storage)
Infrared spectrum Conforms
Suitability (for Fe determination) Passes test
UV at 450 nm A: ≤0.01
Color scale ≤20 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR