Pentafluoropropionic acid 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Perfluoropropionic acid, Pentafluoropropanoic acid, Perfluoropropanoic acid, 2,2,3,3,3-Pentafluoropropionic acid

416920100 416920500 416922500
ACRO416920100EA 24.8 GBP
ACRO416920100 ACRO416920500 ACRO416922500
Pentafluoropropionic acid 97%
Pentafluoropropionic acid
Formula: C₃HF₅O₂
MW: 164.03 g/mol
Boiling Pt: 96 °C (760 mmHg)
Density: 1.576 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004170
CAS Number: 422-64-0
EINECS: 207-021-6
UN: 3265
ADR: 8 III

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR