Peanut oil

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

416855000 416850025
ACRO416855000EA 110 GBP
ACRO416855000 ACRO416850025
Peanut oil
Peanut oil
Density: 0.915 g/cm³
Flash Pt: 283 °C
Storage Temperature: Ambient
MDL Number: MFCD00131805
CAS Number: 8002-03-7

Order Now

Specification Test Results

Appearance Clear yellow Oily liquid
Infrared spectrum Conforms
Water ≤0.1 %
Composition of fatty acids 0.5 to 3.0 % (GC) Lignoceric acid
Acid value ≤0.5 mg KOH/g
Peroxide value ≤5 meq O2/kg
Unsaponifiable matter ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR