3,5-Dihydroxytoluene 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Orcinol, 5-Methylbenzene-1,3-diol, 5-Methylresorcinol

416580050 416580250 416581000
ACRO416580050EA 38.8 GBP
ACRO416580050 ACRO416580250 ACRO416581000
3,5-Dihydroxytoluene 98%
3,5-Dihydroxytoluene
Formula: C₇H₈O₂
Boiling Pt: 290 °C (760 mmHg)
Melting Pt: 105…110 °C
Density: 1,2895 g/cm³ (20 °C)
Flash Pt: 159 °C
Storage Temperature: Ambient
MDL Number: MFCD00002291
CAS Number: 504-15-4
EINECS: 207-984-2
Merck Index: 12,06995

Order Now

Specification Test Results

Appearance Pink to grey to light brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 105 to 110 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR