Mithramycin A (from Streptomyces argillaceus) 90%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Plicamycin

415090050 415090010
ACRO415090050EA 367 GBP
ACRO415090050 ACRO415090010
Mithramycin A (from Streptomyces argillaceus) 90%
Mithramycin A

Appearance: Yellow Powder


Soluble in DMSO (100 mg/mL)

Formula: C₅₂H₇₆O₂₄
MW: 1085.1 g/mol
Melting Pt: 180…183 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00135618
CAS Number: 18378-89-7
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Yellow Powder
Solubility (100 mg/ml in DMSO) Clear yellow
HPLC ≥88 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR