Mineral oil (petroleum), pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Petrolatum, liquid, Mineral oil, Paraffin oil, Nujol

415080010 415080060 415080025 415080100
ACRO415080010EA 33.6 GBP
ACRO415080010 ACRO415080060 ACRO415080025 ACRO415080100
Mineral oil (petroleum), pure
Mineral oil (petroleum)
Density: 0.877 g/cm³
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8042-47-5

Order Now

Specification Test Results

Appearance Clear colorless Oily liquid
Infrared spectrum Conforms
Refractive index 1.4730 to 1.4800 (20°C, 589 nm)
Density (15°C) 0.860 to 0.880 g/ml
Viscosity 63 to 80 mm²/s (40°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR