2-Methoxy-p-cresol 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

413850250 413851000
ACRO413850250EA 37.8 GBP
ACRO413850250 ACRO413851000
2-Methoxy-p-cresol 99%
2-Methoxy-p-cresol
Formula: C₈H₁₀O₂
Boiling Pt: 219…222 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1.09 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00002378
CAS Number: 93-51-6
EINECS: 202-252-9
Merck Index: 13,02599

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid (May turn yellow on storage)
Infrared spectrum Conforms
Refractive index 1.5350 to 1.5400 (20°C, 589 nm)
GC ≥98.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR