2,4-Xylenol 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

408450250 408451000 408450050 408455000
ACRO408450250EA 22.3 GBP
ACRO408450250 ACRO408451000 ACRO408450050 ACRO408455000
2,4-Xylenol 99%
2,4-Xylenol
Formula: C₈H₁₀O
Boiling Pt: 211 °C
Melting Pt: 21…23 °C
Density: 1.027 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00002233
CAS Number: 105-67-9
EINECS: 203-321-6
UN: 2928
ADR: 6.1,II
Merck Index: 12,10215

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5375 to 1.5405 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR