Boron trifluoride - methanol complex 12% 1.5 M in methanol

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

402761000 402765000 402760010
ACRO402761000EA 20.8 GBP
ACRO402761000 ACRO402765000 ACRO402760010
Boron trifluoride - methanol complex 12% 1.5 M in methanol
Boron trifluoride methanol complex
Formula: CH₄BF₃O
MW: 99.85 g/mol
Melting Pt: –98 °C
Density: 0.87 g/cm³
Flash Pt: 4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00071635
CAS Number: 373-57-9
EINECS: 206-766-4
UN: 3286
ADR: 3 II

Order Now

Specification Test Results

Appearance Colorless to amber Solution
Titration with NaOH 10 to 15 % w/v
Refractive index 1.5250 to 1.5300 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR