2'-Aminoacetophenone 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

400620100 400620050 400620500
ACRO400620100EA 42.9 GBP
ACRO400620100 ACRO400620050 ACRO400620500
2'-Aminoacetophenone 97%
2'-Aminoacetophenone
Formula: C₈H₉NO
Boiling Pt: 70…71 °C (3 mmHg)
Melting Pt: 20 °C
Density: 1,12 g/cm³ (20 °C)
Flash Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007717
CAS Number: 551-93-9
EINECS: 209-002-8
Merck Index: 13,00412

Order Now

Specification Test Results

Appearance Yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.6130 to 1.6170 (20°C, 589 nm)
GC ≥96.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR