Agar, powder, pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Agar Agar

400405000 400402500 400400010 400400050 400400250
ACRO400405000EA 106 GBP
ACRO400405000 ACRO400402500 ACRO400400010 ACRO400400050 ACRO400400250
Agar, powder, pure
Agar
Melting Pt: 90
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
EINECS: 232-658-1
Merck Index: 13,00184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to cream to light yellow to beige Powder
Infrared spectrum Conforms
Loss on drying ≤20.0 % (1 g, 105°C, 5 h)
Heavy metals ≤20 ppm
Gel strength 500 to 1000 g/cm² (1.5 % at 20°C)
Ash ≤6 %
Microbial contamination Absent (Salmonella)
pH 5.7 to 7.5 (1.5 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR