N-Succinimidyl Acrylate 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: N-Acryloxysuccinimide, 2,5-Dioxopyrrolidin-1-yl acrylate, N-Acryloyloxysuccinimide, Acrylic Acid N-Succinimidyl Ester

400300050 400300010 400300250
ACRO400300050EA 242 GBP
ACRO400300050 ACRO400300010 ACRO400300250
N-Succinimidyl Acrylate 99%
N-Succinimidyl Acrylate
Formula: C₇H₇NO₄
MW: 169.14 g/mol
Melting Pt: 69 °C
Storage Temperature: Freezer
MDL Number: MFCD00078261
CAS Number: 38862-24-7

Order Now

Specification Test Results

Appearance White to yellow Powder or crystals and/or chunks
Infrared spectrum Conforms
Titration with TBAH ≥98.5 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR