2-Bromo-5-formylpyridine 95%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 6-Bromonicotinaldehyde, 2-Bromopyridine-5-carbaldehyde, 2-Bromopyridine-5-carboxaldehyde, 6-Bromo-3-pyridinecarboxaldehyde

399850010 399850050 399850250
ACRO399850010EA 54.7 GBP
ACRO399850010 ACRO399850050 ACRO399850250
2-Bromo-5-formylpyridine 95%
2-Bromo-5-formylpyridine
Formula: C₆H₄BrNO
Melting Pt: 104…110 °C
Storage Temperature: Ambient
MDL Number: MFCD04115419
CAS Number: 149806-06-4

Order Now

Specification Test Results

Appearance White to brown Crystalline powder
Infrared spectrum Conforms
Melting point 98 to 108 °C
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR