2-Bromo-6-nitrotoluene 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

398420050
ACRO398420050EA 26.4 GBP
ACRO398420050
2-Bromo-6-nitrotoluene 98%
2-Bromo-6-nitrotoluene
Formula: C₇H₆BrNO₂
Boiling Pt: 143 °C (22 mmHg)
Melting Pt: 38…41 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00009792
CAS Number: 55289-35-5

Order Now

Specification Test Results

Appearance Light beige to brown Low melting solid
Infrared spectrum Conforms
Melting point 38 to 42 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR