3-Bromobenzoyl chloride 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: m-Bromobenzoyl chloride, 3-Bromobenzoylchloride

397450050 397450250 397451000
ACRO397450050EA 22.4 GBP
ACRO397450050 ACRO397450250 ACRO397451000
3-Bromobenzoyl chloride 97%
3-Bromobenzoyl chloride
Formula: C₇H₄BrClO
MW: 219.46 g/mol
Boiling Pt: 74.5…75 °C (0.5 mmHg)
Density: 1.666 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00000669
CAS Number: 1711-09-7
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5940 to 1.5960 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR