Potassium acetate 99+% for molecular biology

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

397120010 397121000 397122500
ACRO397120010EA 122 GBP
ACRO397120010 ACRO397121000 ACRO397122500
Potassium acetate 99+% for molecular biology
Potassium acetate
Formula: CH₃COOK
Melting Pt: 292 °C
Density: 1,57 g/cm³ (25 °C)
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Powder or crystals
Infrared spectrum Conforms
Loss on drying ≤2 %
Titration with HClO4 ≥99.0 % (On dry substance)
Dnase,Rnase,Protease activity None detected
Heavy metals (as Pb) ≤4 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR