Sodium-1-decanesulphonate, anhydrous 99+% for ion pair chromatography

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Sodium decane-1-sulphonate, Decane-1-sulphonic acid sodium salt, 1-Decanesulphonic acid, sodium salt

396710250 396711000
ACRO396710250EA 135 GBP
ACRO396710250 ACRO396711000
Sodium-1-decanesulphonate, anhydrous 99+% for ion pair chromatography
Sodium 1-decanesulphonate
Formula: C₁₀H₂₁NaO₃S
MW: 244.33 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007526
CAS Number: 13419-61-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (150°C)
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.2 M soln.) Passes test
Suitability (HPLC gradient) Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR