Cyclopentyl methyl ether (CPME) 99.9+% stabilised, extra pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

396130010 396130025 396130500 396132500
ACRO396130010EA 88.8 GBP
ACRO396130010 ACRO396130025 ACRO396130500 ACRO396132500
Cyclopentyl methyl ether (CPME) 99.9+% stabilised, extra pure
Cyclopentyl methyl ether (CPME)
Formula: C₆H₁₂O
MW: 100.16 g/mol
Melting Pt: –140 °C
Density: 0.86 g/cm³
Flash Pt: –1 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD08276401
CAS Number: 5614-37-9
UN: 3271
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4220 (20°C, 589 nm)
Water ≤0.02 %
GC ≥99.90 %
Color scale ≤10 APHA
Stabilizer 45 to 100 ppm BHT
Residue after evaporation ≤0.01 %
Peroxides ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR