Benzylphosphonic acid 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

393340010 393340050 393340250
ACRO393340010EA 25.7 GBP
ACRO393340010 ACRO393340050 ACRO393340250
Benzylphosphonic acid 98%
Benzylphosphonic acid
Formula: C₇H₉O₃P
Melting Pt: 165…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00039519
CAS Number: 6881-57-8
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 169 to 173 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR