DL-Alanine methyl ester hydrochloride 98+%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: H-DL-Ala-OMe.HCl, H-DL-Ala-OMe·HCl

393310010 393310050 393310250 393311000
ACRO393310010EA 16.6 GBP
ACRO393310010 ACRO393310050 ACRO393310250 ACRO393311000
DL-Alanine methyl ester hydrochloride 98+%
DL-Alanine methyl ester hydrochloride
Formula: C₄H₉NO₂
Melting Pt: 157…158 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00035523
CAS Number: 13515-97-4

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR