Sodium-4-hydroxybenzoate 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

391520250 391521000
ACRO391520250EA 44.7 GBP
ACRO391520250 ACRO391521000
Sodium-4-hydroxybenzoate 98%
Sodium-4-hydroxybenzoate

Appearance: White to light beige Crystalline powder

Formula: C₇H₆O₃
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00016530
CAS Number: 114-63-6

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR