Pumice stone, granular (∼ 1-5 mm)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

390040010 390042500
ACRO390040010EA 54.8 GBP
ACRO390040010 ACRO390042500
Pumice stone, granular (∼ 1-5 mm)
Pumice stone
Storage Temperature: Ambient
MDL Number: MFCD00148147
CAS Number: 1332-09-8

Order Now

Specification Test Results

Appearance Grey-brown Granules
Particle size ≥90 % (0.8 - 3 mm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR