Amberlite® FPC3500 ion-exchange resin

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

389992500 389990010
ACRO389992500EA 28.3 GBP
ACRO389992500 ACRO389990010
Amberlite® FPC3500 ion-exchange resin
Amberlite® FPC3500
Storage Temperature: Ambient
MDL Number: MFCD08460528
CAS Number: 950182-46-4

Order Now

Specification Test Results

Appearance White to light yellow Beads
Particle size ≤5.0 % (> 1.180 mm)
Harmonic mean size 0.450 to 0.650 mm
Uniformity coefficient ≤2.00
Moisture holding capacity 60 to 70 % (H)
Total volume capacity ≥2.60 eq/l (H)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR