Potassium cyanide 97+%, extra pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

388315000 388310025 388310100 388311000
ACRO388315000EA 58.5 GBP
ACRO388315000 ACRO388310025 ACRO388310100 ACRO388311000
Potassium cyanide 97+%, extra pure
Potassium cyanide
Formula: KCN
MW: 65.12 g/mol
Boiling Pt: 1625 °C
Melting Pt: 634 °C
Storage Temperature: Ambient
MDL Number: MFCD00011397
CAS Number: 151-50-8
EINECS: 205-792-3
UN: 1680
ADR: 6.1 I
Merck Index: 13,07709

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Argentometric ≥97.0 %
Water ≤0.2 %
Carbonate (CO3) ≤1 % (as K2CO3)
Potassium formate ≤0.5 %
Free alkali (KOH) ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR