(Trimethylsilyl)diazomethane 2 M in hexanes

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Trimethylsilyldiazomethane, TMS-Diazomethane

385330050 385330250 385331000 385335000
ACRO385330050EA 58.3 GBP
ACRO385330050 ACRO385330250 ACRO385331000 ACRO385335000
(Trimethylsilyl)diazomethane 2 M in hexanes
(Trimethylsilyl)diazomethane
Formula: C₄H₁₀N₂Si
MW: 114.22 g/mol
Density: 0.675 g/cm³
Flash Pt: –23 °C
Storage Temperature: Ambient
MDL Number: MFCD00053946
CAS Number: 18107-18-1
UN: 1992
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear yellow Solution
Strength 1.9 to 2.4 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR