6-Chloro-m-toluic acid 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

381970010 381970050
ACRO381970010EA 26.7 GBP
ACRO381970010 ACRO381970050
6-Chloro-m-toluic acid 98%
6-Chloro-m-toluic acid
Formula: C₈H₇ClO₂
Melting Pt: 148…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00045798
CAS Number: 6342-60-5

Order Now

Specification Test Results

Appearance Off-white to beige Powder
Infrared spectrum Conforms
Melting point 148 to 151 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR