tert-Butyl propargyl ether 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

380910010 380910050
ACRO380910010EA 24.5 GBP
ACRO380910010 ACRO380910050
tert-Butyl propargyl ether 97%
tert-Butyl propargyl ether

Appearance: Clear light yellow Liquid

Formula: C₇H₁₂O
Boiling Pt: 119 °C (760 mmHg)
Density: 0.835 g/cm³
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD07368651
CAS Number: 16314-18-4
UN: 3271
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4190 (20°C, 589 nm)
GC ≥96.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR