N,N-Diformylacetamide 80%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Acetamide, N,N-diformyl-

380450010
ACRO380450010EA 25.7 GBP
ACRO380450010
N,N-Diformylacetamide 80%
N,N-Diformylacetamide
Formula: C₄H₅NO₃
MW: 115.088 g/mol
Boiling Pt: 70 °C (15 mmHg)
Storage Temperature: Refrigerator
MDL Number: MFCD00015916
CAS Number: 26944-31-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥75 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR