tert-Butyl vinyl ether 98% stabilised

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

379120100 379120500
ACRO379120100EA 91.4 GBP
ACRO379120100 ACRO379120500
tert-Butyl vinyl ether 98% stabilised
tert-Butyl vinyl ether

Appearance: Clear colorless to light yellow Liquid

Formula: C₆H₁₂O
Boiling Pt: 78 °C (760 mmHg)
Density: 0.762 g/cm³
Flash Pt: –17 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00048246
CAS Number: 926-02-3
UN: 3271
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3970 to 1.3990 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer Approx. 1000 ppm KOH


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR