Benzyl(triphenylphosphoranylidene)acetate 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Benzyl 2-(triphenylphosphoranylidene)acetate

377220050 377220250 377221000
ACRO377220050EA 54.6 GBP
ACRO377220050 ACRO377220250 ACRO377221000
Benzyl(triphenylphosphoranylidene)acetate 97%
Benzyl(triphenylphosphoranylidene)acetate

CAS No.: 15097-38-8

Formula: C₂₇H₂₃O₂P
MW: 410.453 g/mol
Melting Pt: 120…127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191787
CAS Number: 15097-38-8
UN: 3464
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 120 to 127 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR