N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: N-Boc-trans-4-hydroxy-L-proline Methyl Ester, (2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylic acid 1-tert-Butyl 2-Methyl Ester, 1-tert-Butyl 2-Methyl (2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylate, Boc-Hyp-OMe, 1-(tert-Butyl) 2-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate

376250050
ACRO376250050EA 76.8 GBP
ACRO376250050
N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester 97%
N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester

Appearance: White to light beige Crystalline powder

Formula: C₁₁H₁₉NO₅
MW: 245.28 g/mol
Melting Pt: 91…96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00076981
CAS Number: 74844-91-0

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
GC ≥96.0 %
Specific optical rotation -63° to -67° (20°C, 589 nm) (c=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR