(±)-β-Aminophenethyl alcohol 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

374280010 374280050
ACRO374280010EA 24.4 GBP
ACRO374280010 ACRO374280050
(±)-β-Aminophenethyl alcohol 98%
(±)-β-Aminophenethyl alcohol
Formula: C₈H₁₁NO
Boiling Pt: 261 °C (760 mmHg)
Melting Pt: 53…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00130145
CAS Number: 7568-92-5
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR