Ammonium hydrogen carbonate 98%, pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

370930250 370930010 370930050
ACRO370930250EA 18.5 GBP
ACRO370930250 ACRO370930010 ACRO370930050
Ammonium hydrogen carbonate 98%, pure
Ammonium hydrogen carbonate
Formula: NH₄HCO₃
Melting Pt: 105 °C (dec.)
Density: 1,57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012138
CAS Number: 1066-33-7
EINECS: 213-911-5
Merck Index: 13,00497

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HCl ≥97.5 %
Anticaking agent ≤0.55 % (MgCO3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR