Piperonyloyl chloride 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

370790010
ACRO370790010EA 31.7 GBP
ACRO370790010
Piperonyloyl chloride 98%
Piperonyloyl chloride

Appearance: White to yellow to light beige Crystalline powder or crystals and/or chunks

Formula: C₈H₅ClO₃
Melting Pt: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00016904
CAS Number: 25054-53-9
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to yellow to light beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 77 to 81 °C
GC ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR