Iodine monochloride 1 M in dichloromethane

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

370741000 370748000
ACRO370741000EA 49.1 GBP
ACRO370741000 ACRO370748000
Iodine monochloride 1 M in dichloromethane
Iodine monochloride
Formula: ClI
Storage Temperature: Ambient
MDL Number: MFCD00011354
CAS Number: 7790-99-0
EINECS: 232-236-7
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance Orange to red to brown Solution
Titration with Na2S2O3 0.95 to 1.05 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR