Sodium cyanide 98+%, extra pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

370310010 370310050 370310025
ACRO370310010EA 43.4 GBP
ACRO370310010 ACRO370310050 ACRO370310025
Sodium cyanide 98+%, extra pure
Sodium cyanide
Formula: CNNa
Boiling Pt: 1496 °C (1013 hPa)
Melting Pt: 562 °C
Density: 1,595 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003523
CAS Number: 143-33-9
EINECS: 205-599-4
UN: 1689
ADR: 6.1,I
Merck Index: 13,08679

Order Now

Specification Test Results

Appearance White Powder or granular powder or crystals
Titration Argentometric ≥98.0 %
Water ≤0.1 %
Sodium carbonate (Na2CO3) 0.2 to 0.8 %
Sodium hydroxide (NaOH) 0.3 to 0.8 %
Sodium formate 0.1 to 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR