Toluene-p-sulphonic anhydride 95%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 4-Methylbenzenesulphonic anhydride, p-Toluenesulphonic anhydride

367070050 367070250 367071000
ACRO367070050EA 33.8 GBP
ACRO367070050 ACRO367070250 ACRO367071000
Toluene-p-sulphonic anhydride 95%
Toluene-p-sulphonic anhydride
Formula: C₁₄H₁₄O₅S₂
Melting Pt: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00008548
CAS Number: 4124-41-8
EINECS: 223-926-9
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance Off-white to beige to grey Powder or crystals and/or chunks
Infrared spectrum Conforms
Titration by coupling ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR