Coconut oil, pure, refined

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

365471000 365475000
ACRO365471000EA 17.6 GBP
ACRO365471000 ACRO365475000
Coconut oil, pure, refined
Coconut oil
Melting Pt: 23…26 °C
Flash Pt: >200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00130817
CAS Number: 8001-31-8
EINECS: 232-282-8

Order Now

Specification Test Results

Appearance Colorless to white to light yellow Oily liquid or solid
Acid value ≤0.5 mg KOH/g
Iodine value 7 to 12 g I2/100 g
Peroxide value ≤5 meq O2/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR