Silica gel, functionalized, Amino-3 ∼1.4 mmol/g, Ø 40-63 micron

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

360180100 360180250 360181000
ACRO360180100EA 39.5 GBP
ACRO360180100 ACRO360180250 ACRO360181000
Silica gel, functionalized, Amino-3 ∼1.4 mmol/g, Ø 40-63 micron
Silica gel, functionalized, Amino-3

CAS No.: 71888-97-6

Formula: C₉H₂₃NO₅Si₂
Storage Temperature: Ambient
MDL Number: MFCD03427643
CAS Number: 71888-97-6

Order Now

Specification Test Results

Appearance White to off-white Powder
Particle size 40 to 63 µm
Loading ≥1.2 mmol/g (molecular)
Nitrogen (N) ≥1.68 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR